Assay Details
Gene Symbol: Arfip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004222.1 | NP_001004222.1 | 5 - 6 | 670 | 132 | |
XM_006229933.2 | XP_006229995.1 | 5 - 6 | 688 | 132 | ||
GenBank mRNA | BC079174.1 | - | 5 - 6 | 670 | 7130075 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
