Assay Details
Gene Symbol: Dkk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138519.2 | NP_612528.2 | 3 - 4 | 492 | 125 | |
XM_006230009.2 | XP_006230071.1 | 3 - 4 | 500 | 125 | ||
GenBank mRNA | AF245040.1 | - | 2 - 3 | 358 | 125 | |
BC085702.1 | - | 2 - 3 | 352 | 125 | ||
BC160821.1 | - | 3 - 4 | 492 | 7321198 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
