Assay Details
Gene Symbol: Clns1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031719.1 | NP_113907.1 | 3 - 4 | 406 | 71 | |
XM_006229792.2 | XP_006229854.1 | 3 - 4 | 469 | 71 | ||
GenBank mRNA | D13985.1 | - | 3 - 4 | 476 | 71 | |
D17698.1 | - | 3 - 4 | 389 | 71 | ||
FQ212666.1 | - | 1 - 2 | 88 | 71 | ||
FQ232880.1 | - | 1 - 2 | 88 | 71 | ||
L26450.1 | - | 3 - 4 | 406 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
