Assay Details
Gene Symbol: Clns1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229792.2 | XP_006229854.1 | 6 - 7 | 845 | 75 | |
GenBank mRNA | D13985.1 | - | 6 - 7 | 852 | 75 | |
FQ212666.1 | - | 4 - 5 | 464 | 75 | ||
FQ232880.1 | - | 4 - 5 | 465 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
