Assay Details
Gene Symbol: Ate1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106300.1 | NP_001099770.1 | 1414 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P527376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919621 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937371 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953794 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903471 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357367 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796790 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170008 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792466 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819411 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93965 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53226 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103872 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P527376 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P527376 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P527376 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1052798 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P527376 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 102116 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 26267 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 1067 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 72756 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37914 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10726206 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10726207 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10726194 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10726198 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10726204 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10726202 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10726196 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10726195 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10726205 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10726203 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10726201 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10726197 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10726193 | - | - |
Rat Genome 230 2.0 Array | 1377820_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1394118_at | - | - |
Rat Genome 230 2.0 Array | 1394118_PM_at | - | - |
Rat Genome 230 2.0 Array | 1393296_PM_at | - | - |
Rat Genome 230 2.0 Array | 1377820_a_at | - | - |
Rat Genome 230 2.0 Array | 1393296_at | - | - |
Affymetrix HT Rat Focus Array | 1377820_a_at | - | - |
Affymetrix HT Rat Focus Array | 1393296_at | - | - |
Affymetrix HT Rat Focus Array | 1377820_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1394118_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1394118_at | - | - |
Affymetrix HT Rat Focus Array | 1393296_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100053335.rn.1 | TC0100006754.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100053336.rn.1 | TC0100006754.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100053337.rn.1 | TC0100006754.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100053338.rn.1 | TC0100006754.rn.1 | RTA TC |
Rat Expression Set 230 | 1377820_a_at | - | - |
Rat Expression Set 230 | 1394118_PM_at | - | - |
Rat Expression Set 230 | 1394118_at | - | - |
Rat Expression Set 230 | 1377820_PM_a_at | - | - |
Rat Expression Set 230 | 1393296_PM_at | - | - |
Rat Expression Set 230 | 1393296_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10726193 | - | - |
Rat Genome U34 Set | rc_AA894340_at | - | - |
Rat Genome U34 Set | rc_AI010248_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21136075 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359229 | - | - |
