Assay Details
Gene Symbol: Tufm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106295.1 | NP_001099765.1 | 3 - 4 | 742 | 77 | |
XM_006230224.1 | XP_006230286.1 | 3 - 4 | 737 | 77 | ||
XM_006230225.1 | XP_006230287.1 | 3 - 4 | 737 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
