Assay Details
Gene Symbol: Tmbim4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199116.2 | NP_954547.1 | 4 - 5 | 406 | 96 | |
GenBank mRNA | BC060596.1 | - | 4 - 5 | 407 | 96 | |
FQ217672.1 | - | 4 - 5 | 349 | 96 | ||
FQ228009.1 | - | 4 - 5 | 403 | 96 | ||
FQ233689.1 | - | 4 - 5 | 400 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
