Assay Details
Gene Symbol: Tcp11l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017458.1 | NP_001017458.1 | 1 - 2 | 222 | 80 | |
XM_006241174.2 | XP_006241236.1 | 2 - 3 | 284 | 80 | ||
GenBank mRNA | BC092644.1 | - | 1 - 2 | 222 | 7318345 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
