Assay Details
Gene Symbol: Tcp11l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017458.1 | NP_001017458.1 | 2 - 3 | 358 | 137 | |
XM_006241174.2 | XP_006241236.1 | 3 - 4 | 420 | 137 | ||
GenBank mRNA | BC092644.1 | - | 2 - 3 | 358 | 137 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
