Assay Details
Gene Symbol: Tcp11l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017458.1 | NP_001017458.1 | 4 - 5 | 703 | 71 | |
XM_006241174.2 | XP_006241236.1 | 5 - 6 | 765 | 71 | ||
GenBank mRNA | BC092644.1 | - | 4 - 5 | 703 | 7318345 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
