Assay Details
Gene Symbol: Metap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022539.1 | NP_071984.1 | 6 - 7 | 944 | 154 | |
GenBank mRNA | BC070892.1 | - | 6 - 7 | 859 | 154 | |
BC091150.1 | - | 6 - 7 | 958 | 154 | ||
L10652.1 | - | 6 - 7 | 944 | 154 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
