Assay Details
Gene Symbol: Chpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007750.1 | NP_001007751.1 | 3 - 4 | 662 | 80 | |
XM_006241228.2 | XP_006241290.1 | 3 - 4 | 657 | 80 | ||
XM_006241229.2 | XP_006241291.1 | 3 - 4 | 657 | 80 | ||
XM_006241230.2 | XP_006241292.1 | 3 - 4 | 657 | 80 | ||
XM_006241231.2 | XP_006241293.1 | 3 - 4 | 657 | 80 | ||
GenBank mRNA | BC082074.1 | - | 3 - 4 | 662 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
