Assay Details
Gene Symbol: Csrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177425.3 | NP_803174.2 | 3 - 4 | 330 | 129 | |
GenBank mRNA | BC086356.1 | - | 3 - 4 | 344 | 129 | |
CD373038.1 | - | 3 - 4 | 321 | 129 | ||
U44948.1 | - | 3 - 4 | 335 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
