Assay Details
Gene Symbol: Elk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108743.1 | NP_001102213.1 | 2 - 3 | 1007 | 58 | |
XM_006241265.2 | XP_006241327.1 | 3 - 4 | 1412 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
