Assay Details
Gene Symbol: Elk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108743.1 | NP_001102213.1 | 3 - 4 | 1130 | 66 | |
XM_006241265.2 | XP_006241327.1 | 4 - 5 | 1535 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
regulation of transcription, DNA-templated
regulation of transcription by RNA polymerase II
transcription by RNA polymerase II
cell differentiation
wound healing
negative regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
RNA polymerase II cis-regulatory region sequence-specific DNA binding
DNA-binding transcription factor activity, RNA polymerase II-specific
DNA binding
DNA-binding transcription factor activity
transcription corepressor activity
purine-rich negative regulatory element binding
angiogenesis
regulation of transcription, DNA-templated
regulation of transcription by RNA polymerase II
transcription by RNA polymerase II
cell differentiation
wound healing
negative regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
RNA polymerase II cis-regulatory region sequence-specific DNA binding
DNA-binding transcription factor activity, RNA polymerase II-specific
DNA binding
DNA-binding transcription factor activity
transcription corepressor activity
purine-rich negative regulatory element binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P178811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250010 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872335 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749963 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759221 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43507 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102362 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P178811 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P178811 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P178811 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P178811 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P199447 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 28831 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10901908 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10901907 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10901905 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10901906 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10901904 | - | - |
Rat Genome 230 2.0 Array | 1389617_PM_at | - | - |
Rat Genome 230 2.0 Array | 1394473_PM_at | - | - |
Rat Genome 230 2.0 Array | 1394473_at | - | - |
Rat Genome 230 2.0 Array | 1384306_PM_at | - | - |
Rat Genome 230 2.0 Array | 1374134_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389617_at | - | - |
Rat Genome 230 2.0 Array | 1374134_at | - | - |
Rat Genome 230 2.0 Array | 1384306_at | - | - |
Affymetrix HT Rat Focus Array | 1374134_at | - | - |
Affymetrix HT Rat Focus Array | 1384306_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1374134_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1384306_at | - | - |
Affymetrix HT Rat Focus Array | 1389617_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1394473_at | - | - |
Affymetrix HT Rat Focus Array | 1389617_at | - | - |
Affymetrix HT Rat Focus Array | 1394473_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700019918.rn.1 | TC0700002491.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700019917.rn.1 | TC0700002491.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700019914.rn.1 | TC0700002491.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700019915.rn.1 | TC0700002491.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700019916.rn.1 | TC0700002491.rn.1 | RTA TC |
Rat Expression Set 230 | 1374134_at | - | - |
Rat Expression Set 230 | 1389617_PM_at | - | - |
Rat Expression Set 230 | 1374134_PM_at | - | - |
Rat Expression Set 230 | 1389617_at | - | - |
Rat Expression Set 230 | 1394473_PM_at | - | - |
Rat Expression Set 230 | 1384306_at | - | - |
Rat Expression Set 230 | 1384306_PM_at | - | - |
Rat Expression Set 230 | 1394473_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10901904 | - | - |
Rat Genome U34 Set | rc_AA925514_at | - | - |
Rat Genome U34 Set | rc_AA900762_at | - | - |
Rat Genome U34 Set | rc_AI070663_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21180296 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373250 | - | - |
