Assay Details
Gene Symbol: Slc25a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139100.2 | NP_620800.2 | 3 - 4 | 371 | 128 | |
GenBank mRNA | CB577607.1 | - | 3 - 4 | 371 | 128 | |
FQ210190.1 | - | 3 - 4 | 325 | 128 | ||
FQ213750.1 | - | 3 - 4 | 330 | 128 | ||
FQ220537.1 | - | 3 - 4 | 325 | 128 | ||
FQ220626.1 | - | 3 - 4 | 312 | 128 | ||
FQ227446.1 | - | 3 - 4 | 325 | 128 | ||
FQ229113.1 | - | 3 - 4 | 325 | 128 | ||
FQ230627.1 | - | 3 - 4 | 325 | 128 | ||
FQ232293.1 | - | 1 - 2 | 130 | 128 | ||
FQ232835.1 | - | 3 - 4 | 321 | 128 | ||
FQ234854.1 | - | 3 - 4 | 325 | 128 | ||
M23984.1 | - | 2 - 3 | 290 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
