Assay Details
Gene Symbol: Helb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241426.2 | XP_006241488.1 | 5 - 6 | 2189 | 69 | |
XM_006241428.2 | XP_006241490.1 | 4 - 5 | 1942 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
