Assay Details
Gene Symbol: Actr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108081.1 | NP_001101551.1 | 1 - 2 | 121 | 143 | |
GenBank mRNA | BC166860.1 | - | 1 - 2 | 97 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
