Assay Details
Gene Symbol: Actr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108081.1 | NP_001101551.1 | 2 - 3 | 246 | 77 | |
XM_006241243.1 | XP_006241305.1 | 2 - 3 | 255 | 77 | ||
GenBank mRNA | BC166860.1 | - | 2 - 3 | 222 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
