Assay Details
Gene Symbol: Actr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108081.1 | NP_001101551.1 | 4 - 5 | 438 | 127 | |
XM_006241243.1 | XP_006241305.1 | 4 - 5 | 447 | 127 | ||
GenBank mRNA | BC166860.1 | - | 4 - 5 | 414 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
