Assay Details
Gene Symbol: Actr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108081.1 | NP_001101551.1 | 7 - 8 | 721 | 96 | |
XM_006241243.1 | XP_006241305.1 | 7 - 8 | 730 | 96 | ||
GenBank mRNA | BC166860.1 | - | 7 - 8 | 697 | 96 | |
FQ220136.1 | - | 4 - 5 | 366 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
