Assay Details
Gene Symbol: Timp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012886.2 | NP_037018.2 | 3 - 4 | 607 | 76 | |
XM_006241162.2 | XP_006241224.1 | 3 - 4 | 636 | 76 | ||
GenBank mRNA | BC097335.1 | - | 3 - 4 | 607 | 76 | |
U27201.1 | - | 3 - 4 | 322 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
