Assay Details
Gene Symbol: Msrb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001055430.5 | XP_001055430.4 | 3 - 4 | 841 | 66 | |
XM_006241414.2 | XP_006241476.1 | 4 - 5 | 435 | 66 | ||
XM_008765427.1 | XP_008763649.1 | 3 - 4 | 861 | 66 | ||
XM_008765428.1 | XP_008763650.1 | 4 - 5 | 485 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
