Assay Details
Gene Symbol: Msrb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001055430.5 | XP_001055430.4 | 6 - 7 | 1043 | 103 | |
XM_006241414.2 | XP_006241476.1 | 7 - 8 | 637 | 103 | ||
XM_008765427.1 | XP_008763649.1 | 6 - 7 | 1063 | 103 | ||
XM_008765428.1 | XP_008763650.1 | 7 - 8 | 687 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
