Assay Details
Gene Symbol: Pawr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033485.2 | NP_277020.1 | 5 - 6 | 974 | 143 | |
XM_006241317.2 | XP_006241379.1 | 6 - 7 | 1187 | 143 | ||
GenBank mRNA | U05989.1 | - | 5 - 6 | 974 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
negative regulation of B cell proliferation
negative regulation of T cell proliferation
positive regulation of amyloid precursor protein biosynthetic process
negative regulation of T cell receptor signaling pathway
actin filament bundle assembly
apoptotic signaling pathway
actin binding
protein binding
enzyme binding
leucine zipper domain binding
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
negative regulation of B cell proliferation
negative regulation of T cell proliferation
positive regulation of amyloid precursor protein biosynthetic process
negative regulation of T cell receptor signaling pathway
actin filament bundle assembly
apoptotic signaling pathway
actin binding
protein binding
enzyme binding
leucine zipper domain binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P12739 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930918 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556269 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736182 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933556 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23116 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P12739 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P12739 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P12739 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12739 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700936259 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702156525 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32498 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71841 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 76006 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 9473 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 92087 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 20260 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 18287 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 702156525 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700936259 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10895340 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895338 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895341 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895343 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895339 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895342 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10895337 | - | - |
Rat Genome 230 2.0 Array | 1368702_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368702_at | - | - |
Affymetrix HT Rat Focus Array | 1368702_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368702_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700020751.rn.1 | TC0700002661.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700020752.rn.1 | TC0700002661.rn.1 | RTA TC |
Rat Expression Set 230 | 1368702_PM_at | - | - |
Rat Expression Set 230 | 1368702_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10895337 | - | - |
Rat Genome U34 Set | U05989_at | - | - |
Rat Genome U34 Set | U05989_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22164269 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369640 | - | - |
RatRef-12 Expression BeadChip | ILMN_56974 | - | - |
