Assay Details
Gene Symbol: P2rx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270621.1 | NP_001257550.1 | 10 - 11 | 1185 | 89 | |
NM_031075.2 | NP_112337.2 | 11 - 12 | 1257 | 89 | ||
NR_073054.1 | 12 - 13 | 1279 | 89 | |||
GenBank mRNA | AF084975.1 | - | 10 - 11 | 1030 | 89 | |
BC081783.1 | - | 12 - 13 | 1279 | 89 | ||
X90651.1 | - | 11 - 12 | 1257 | 89 | ||
X91167.1 | - | 11 - 12 | 1247 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
