Assay Details
Gene Symbol: P2rx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270621.1 | NP_001257550.1 | 11 | 2842 | 115 | |
NM_031075.2 | NP_112337.2 | 12 | 2914 | 115 | ||
NR_073054.1 | 13 | 2936 | 115 | |||
GenBank mRNA | X90651.1 | - | 12 | 2893 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
ion transport
cation transport
chemical synaptic transmission
neuromuscular synaptic transmission
response to temperature stimulus
response to heat
response to cold
response to mechanical stimulus
response to carbohydrate
response to organic substance
urinary bladder smooth muscle contraction
neuronal action potential
peristalsis
response to ATP
purinergic nucleotide receptor signaling pathway
regulation of synaptic plasticity
behavioral response to pain
sensory perception of taste
protein homooligomerization
cation transmembrane transport
purinergic nucleotide receptor activity
extracellularly ATP-gated cation channel activity
ATP binding
response to hypoxia
ion transport
cation transport
chemical synaptic transmission
neuromuscular synaptic transmission
response to temperature stimulus
response to heat
response to cold
response to mechanical stimulus
response to carbohydrate
response to organic substance
urinary bladder smooth muscle contraction
neuronal action potential
peristalsis
response to ATP
purinergic nucleotide receptor signaling pathway
regulation of synaptic plasticity
behavioral response to pain
sensory perception of taste
protein homooligomerization
cation transmembrane transport
purinergic nucleotide receptor activity
extracellularly ATP-gated cation channel activity
ATP binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P018681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278282 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838969 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268764 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118757 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P018681 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P018681 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P018681 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P15314 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P198746 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 10429 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 44534 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 22794 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 56009 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 77285 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10846896 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10846897 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10846888 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10846892 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10846891 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10846889 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10846882 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10846884 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10846895 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10846898 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10846890 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10846886 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10846883 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10846894 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10846899 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10846900 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10846885 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10846887 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10846881 | - | - |
Rat Genome 230 2.0 Array | 1368667_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368667_at | - | - |
Rat Genome 230 2.0 Array | 1388077_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1388077_a_at | - | - |
Affymetrix HT Rat Focus Array | 1388077_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1388077_a_at | - | - |
Affymetrix HT Rat Focus Array | 1368667_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368667_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300027169.rn.1 | TC0300003447.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300027173.rn.1 | TC0300003447.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300027170.rn.1 | TC0300003447.rn.1 | RTA TC |
Rat Expression Set 230 | 1368667_at | - | - |
Rat Expression Set 230 | 1388077_a_at | - | - |
Rat Expression Set 230 | 1388077_PM_a_at | - | - |
Rat Expression Set 230 | 1368667_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10846881 | - | - |
Rat Genome U34 Set | X90651_at | - | - |
Rat Genome U34 Set | X90651_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21386266 | - | - |
