Assay Details
Gene Symbol: Folh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057185.2 | NP_476533.1 | 17 - 18 | 2042 | 90 | |
GenBank mRNA | AF039707.1 | - | 11 - 12 | 1128 | 90 | |
AF040256.1 | - | 17 - 18 | 2048 | 90 | ||
AF513486.2 | - | 17 - 18 | 2042 | 90 | ||
U75973.1 | - | 17 - 18 | 1999 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
