Assay Details
Gene Symbol: Arl6ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB190510.1 | - | 2 - 3 | 247 | 85 |
Gene Symbol: Smg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759828.1 | XP_008758050.1 | 59 - 60 | 10941 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
