Assay Details
Gene Symbol: Eif3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100662.1 | NP_001094132.1 | 16 - 17 | 2386 | 99 | |
XM_006230226.2 | XP_006230288.1 | 16 - 17 | 2702 | 99 | ||
GenBank mRNA | BC103728.1 | - | 10 - 11 | 1576 | 99 | |
BC169011.1 | - | 16 - 17 | 2386 | 9090822 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
