Assay Details
Gene Symbol: Eif3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100662.1 | NP_001094132.1 | 1 - 2 | 167 | 67 | |
XM_006230226.2 | XP_006230288.1 | 1 - 2 | 483 | 67 | ||
GenBank mRNA | BC169011.1 | - | 1 - 2 | 167 | 9090822 | 67 |
FQ225044.1 | - | 1 - 2 | 270 | 67 | ||
FQ227337.1 | - | 1 - 2 | 270 | 67 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC169011.1,NM_001100662.1,XM_006230226.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
