Assay Details
Gene Symbol: Palb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191603.1 | NP_001178532.1 | 7 - 8 | 2524 | 108 | |
XM_008759711.1 | XP_008757933.1 | 5 - 6 | 3700 | 108 | ||
XM_008759712.1 | XP_008757934.1 | 6 - 7 | 2608 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
