Assay Details
Gene Symbol: Eif3f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277302.1 | NP_001264231.1 | 7 - 8 | 1035 | 116 | |
GenBank mRNA | FQ213714.1 | - | 7 - 8 | 1036 | 116 | |
FQ219431.1 | - | 2 - 3 | 228 | 116 | ||
FQ229379.1 | - | 7 - 8 | 709 | 116 | ||
FQ230130.1 | - | 7 - 8 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
