Assay Details
Gene Symbol: Rnf121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107540.2 | NP_001101010.1 | 7 - 8 | 756 | 72 | |
XM_006229878.2 | XP_006229940.1 | 7 - 8 | 981 | 72 | ||
XM_006229879.2 | XP_006229941.1 | 8 - 9 | 1099 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
