Assay Details
Gene Symbol: Rnf121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107540.2 | NP_001101010.1 | 8 - 9 | 863 | 99 | |
XM_006229878.2 | XP_006229940.1 | 8 - 9 | 1088 | 99 | ||
XM_006229879.2 | XP_006229941.1 | 9 - 10 | 1206 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P002076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739431 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400307 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971371 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118016 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103063 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P002076 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P002076 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P002076 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P510918 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 31644 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 41999 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10724149 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10724140 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10724148 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10724139 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10724145 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10724142 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10724143 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10724147 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10724144 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10724146 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10724141 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10724138 | - | - |
Rat Genome 230 2.0 Array | 1385644_PM_at | - | - |
Rat Genome 230 2.0 Array | 1385644_at | - | - |
Rat Genome 230 2.0 Array | 1386639_at | - | - |
Rat Genome 230 2.0 Array | 1385403_at | - | - |
Rat Genome 230 2.0 Array | 1386639_PM_at | - | - |
Rat Genome 230 2.0 Array | 1385403_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1385644_at | - | - |
Affymetrix HT Rat Focus Array | 1385644_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1386639_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1385403_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1385403_at | - | - |
Affymetrix HT Rat Focus Array | 1386639_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100049112.rn.1 | TC0100006174.rn.1 | RTA TC |
Rat Expression Set 230 | 1385403_at | - | - |
Rat Expression Set 230 | 1385644_PM_at | - | - |
Rat Expression Set 230 | 1386639_PM_at | - | - |
Rat Expression Set 230 | 1385403_PM_at | - | - |
Rat Expression Set 230 | 1385644_at | - | - |
Rat Expression Set 230 | 1386639_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10724138 | - | - |
Rat Genome U34 Set | rc_AI058291_at | - | - |
Rat Genome U34 Set | rc_AI071834_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364199 | - | - |
