Assay Details
Gene Symbol: Rnf121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229879.2 | XP_006229941.1 | 3 - 4 | 442 | 132 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006229879.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
