Assay Details
Gene Symbol: Pold3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024750.1 | NP_001019921.1 | 10 - 11 | 1200 | 140 | |
GenBank mRNA | BC098010.1 | - | 10 - 11 | 1200 | 7454306 | 140 |
FQ211238.1 | - | 1201 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
