Assay Details
Gene Symbol: Pold3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024750.1 | NP_001019921.1 | 2 - 3 | 208 | 85 | |
GenBank mRNA | BC098010.1 | - | 2 - 3 | 208 | 85 | |
FQ211238.1 | - | 210 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
