Assay Details
Gene Symbol: Scube2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748970.3 | XP_003749018.1 | 1 - 2 | 237 | 55 | |
XM_006230006.2 | XP_006230068.1 | 1 - 2 | 237 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
