Assay Details
Gene Symbol: Plk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017100.1 | NP_058796.1 | 9 - 10 | 1704 | 101 | |
GenBank mRNA | BC083926.1 | - | 9 - 10 | 1680 | 7135428 | 101 |
U10188.1 | - | 9 - 10 | 1704 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
