Assay Details
Gene Symbol: Plk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017100.1 | NP_058796.1 | 5 - 6 | 1137 | 101 | |
GenBank mRNA | BC083926.1 | - | 5 - 6 | 1113 | 101 | |
U10188.1 | - | 5 - 6 | 1137 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
