Assay Details
Gene Symbol: Ilk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133409.2 | NP_596900.1 | 9 - 10 | 1036 | 164 | |
GenBank mRNA | AF329194.1 | - | 9 - 10 | 973 | 164 | |
BC062406.1 | - | 9 - 10 | 1036 | 5623251 | 164 | |
FQ233927.1 | - | 3 - 4 | 363 | 164 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
