Assay Details
Gene Symbol: Lcmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199405.2 | NP_955437.1 | 2 - 3 | 311 | 121 | |
XM_006230191.2 | XP_006230253.1 | 2 - 3 | 347 | 121 | ||
GenBank mRNA | BC063186.1 | - | 2 - 3 | 314 | 6890062 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
