Assay Details
Gene Symbol: Lcmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199405.2 | NP_955437.1 | 4 - 5 | 510 | 146 | |
XM_006230191.2 | XP_006230253.1 | 4 - 5 | 546 | 146 | ||
GenBank mRNA | BC063186.1 | - | 4 - 5 | 513 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
