Assay Details
Gene Symbol: Lcmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199405.2 | NP_955437.1 | 8 - 9 | 895 | 147 | |
XM_006230191.2 | XP_006230253.1 | 8 - 9 | 931 | 147 | ||
GenBank mRNA | BC063186.1 | - | 8 - 9 | 898 | 147 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
