Assay Details
Gene Symbol: Psma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017278.1 | NP_058974.1 | 3 - 4 | 239 | 69 | |
GenBank mRNA | BC062233.1 | - | 3 - 4 | 236 | 6919336 | 69 |
D90265.1 | - | 3 - 4 | 239 | 69 | ||
FQ213606.1 | - | 3 - 4 | 229 | 69 | ||
FQ215179.1 | - | 3 - 4 | 218 | 69 | ||
FQ215378.1 | - | 3 - 4 | 222 | 69 | ||
FQ216107.1 | - | 3 - 4 | 232 | 69 | ||
FQ218243.1 | - | 271 | 69 | |||
FQ220925.1 | - | 3 - 4 | 204 | 69 | ||
FQ223714.1 | - | 3 - 4 | 208 | 69 | ||
FQ225726.1 | - | 3 - 4 | 190 | 69 | ||
FQ229288.1 | - | 3 - 4 | 266 | 69 | ||
M29859.1 | - | 3 - 4 | 239 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
