Assay Details
Gene Symbol: Relt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108495.1 | NP_001101965.1 | 1 - 2 | 79 | 74 | |
XM_006229775.2 | XP_006229837.1 | 2 - 3 | 247 | 74 | ||
XM_006229776.2 | XP_006229838.1 | 2 - 3 | 2511 | 74 | ||
XM_006229777.2 | XP_006229839.1 | 2 - 3 | 198 | 74 | ||
XM_006229778.2 | XP_006229840.1 | 2 - 3 | 4599 | 74 | ||
XM_006229780.2 | XP_006229842.1 | 2 - 3 | 247 | 74 | ||
XM_008759763.1 | XP_008757985.1 | 2 - 3 | 4927 | 74 | ||
XM_008759764.1 | XP_008757986.1 | 2 - 3 | 4852 | 74 | ||
XM_008759765.1 | XP_008757987.1 | 2 - 3 | 4807 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
