Assay Details
Gene Symbol: Relt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108495.1 | NP_001101965.1 | 9 - 10 | 1271 | 74 | |
XM_006229775.2 | XP_006229837.1 | 10 - 11 | 1439 | 74 | ||
XM_006229776.2 | XP_006229838.1 | 10 - 11 | 3703 | 74 | ||
XM_006229777.2 | XP_006229839.1 | 10 - 11 | 1390 | 74 | ||
XM_006229778.2 | XP_006229840.1 | 10 - 11 | 5791 | 74 | ||
XM_008759763.1 | XP_008757985.1 | 10 - 11 | 6119 | 74 | ||
XM_008759764.1 | XP_008757986.1 | 10 - 11 | 6044 | 74 | ||
XM_008759765.1 | XP_008757987.1 | 10 - 11 | 5999 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
