Assay Details
Gene Symbol: Pdilt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013902.1 | NP_001013924.1 | 7 - 8 | 1121 | 135 | |
GenBank mRNA | BC083897.1 | - | 7 - 8 | 1121 | 7133354 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
