Assay Details
Gene Symbol: Tgfb1i1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191840.1 | NP_001178769.1 | 7 - 8 | 742 | 102 | |
XM_006230349.2 | XP_006230411.1 | 7 - 8 | 1592 | 102 | ||
XM_006230350.2 | XP_006230412.1 | 7 - 8 | 714 | 102 | ||
XM_008759920.1 | XP_008758142.1 | 6 - 7 | 1799 | 102 | ||
GenBank mRNA | AF314960.1 | - | 3 - 4 | 322 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
